missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567918
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. A range of agonists acting through G protein-coupled receptors and receptor tyrosine kinases stimulate this hydrolysis. PC-specific PLD activity has been implicated in numerous cellular pathways, including signal transduction, membrane trafficking, and the regulation of mitosis (Hammond et al., 1995).
Specifications
| PLD1 | |
| Polyclonal | |
| Unconjugated | |
| PLD1 | |
| AA536939; C85393; choline phosphatase 1; hPLD1; mPLD1; phophatidylcholine-specific; Phoshpolipase D gene 1 probably with two splicing variants A and B; phosphatidylcholine-hydrolyzing phospholipase D1; phospholipase D gene 1, probably with two splicing variants A and B; phospholipase D1; phospholipase D1, phosphatidylcholine-specific; PLD 1; Pld1; Pld1a; Pld1b; Plda; Pldb; rPLD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18805, 25096, 5337 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| P70496, Q13393, Q9Z280 | |
| PLD1 | |
| Synthesized peptide derived from human PLD1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction