Learn More
Invitrogen™ PLD3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA542640
Product Code. 15525592
  • € 518.00 / 100µL
Description
Peptide sequence: WVLLVLILAV VGFGALMTQL FLWEYGDLHL FGPNQRPAPC YDPCEAVLVE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLD3
Polyclonal
Unconjugated
PLD3
5'-3' exonuclease PLD3; phospholipase D3; AD19; Choline phosphatase 3; hindIII K4L homolog; HUK4; HU-K4; phosphatidylcholine-hydrolyzing phospholipase D3; phospholipase D family member 3; phospholipase D family, member 3; phospholipase D3; PLD 3; Pld3; Sam9; Sam-9; Schwannoma-associated protein 9; Unknown (protein for MGC:126910)
Rabbit
Affinity chromatography
RUO
23646
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Surface Plasmon Resonance
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8IV08
PLD3
Synthetic peptide directed towards the N-terminal of human PLD3 (aa 38-87).