missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLDN Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504279
This item is not returnable.
View return policy
Description
The protein encoded by this gene may play a role in intracellular vesicle trafficking. It interacts with Syntaxin 13 which mediates intracellular membrane fusion. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
| PLDN | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9R0C0, Q9UL45 | |
| BLOC1S6 | |
| Full length human recombit protein of human PLDN produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H9 | |
| Unconjugated | |
| BLOC1S6 | |
| BLOS6, HPS9, PA, PALLID, PLDN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18457, 26258 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction