missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLDN Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504279S
This item is not returnable.
View return policy
Description
PLDN Monoclonal antibody specifically detects PLDN in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| PLDN | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14593 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8625 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H9 | |
| Unconjugated | |
| RFXANK | |
| biogenesis of lysosomal organelles complex 1 subunit 6; biogenesis of lysosomal organelles complex-1, subunit 5, pallidin; biogenesis of lysosomal organelles complex-1, subunit 6, pallidin; biogenesis of lysosome-related organelles complex 1 subunit 6; biogenesis of organelles complex-1, subunit 6, pallidin; BLOC-1; BLOC-1 subunit 6; Bloc1s6; BLOS6; HPS9; P2; PA; PALLID; Pallid protein; pallid protein homolog; Pallidin; pallidin homolog; PLDN; Stx13bp1; syntaxin 13 binding protein 1; Syntaxin 13-interacting protein; syntaxin 13-interacting protein pallid | |
| Full length human recombinant protein of human RFXANK produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction