missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pleckstrin Monoclonal Antibody (OTI4G6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525038
This item is not returnable.
View return policy
Description
Pleckstrin Monoclonal Antibody for Western Blot, IHC (P), Flow
Major protein kinase C substrate of platelets.
Specifications
| Pleckstrin | |
| Monoclonal | |
| 0.78 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08567 | |
| PLEK | |
| Full length human recombinant protein of PLEK produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G6 | |
| Unconjugated | |
| PLEK | |
| 2010300B13Rik; FLJ27168; P47; platelet 47 kDa protein; Pleckstrin; Plek | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5341 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction