Learn More
Invitrogen™ Plectin Recombinant Rabbit Monoclonal Antibody (SY29-04)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532102
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Plectin is a prominent member of an important family of structurally and in part functionally related proteins, termed plakins or cytolinkers, that are capable of interlinking different elements of the cytoskeleton. Plakins, with their multi-domain structure and enormous size, not only play crucial roles in maintaining cell and tissue integrity and orchestrating dynamic changes in cytoarchitecture and cell shape, but also serve as scaffolding platforms for the assembly, positioning, and regulation of signaling complexes. Plectin is expressed as several protein isoforms in a wide range of cell types and tissues from a single gene located on chromosome 8 in humans. The expression of specific plectin isoforms was found to be dependent on cell type (tissue) and stage of development and it appears that each cell type (tissue) contains a unique set (proportion and composition) of plectin isoforms which carry out distinct and specific functions.
Specifications
| Plectin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| P30427, Q15149, Q9QXS1 | |
| Plec | |
| Recombinant protein within Human Plectin aa 2-101. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| SY29-04 | |
| Unconjugated | |
| Plec | |
| AA591047; AU042537; EBS1; EBSMD; EBSND; EBSO; EBSOG; EBSPA; HD1; Hemidesmosomal protein 1; LGMD2Q; PCN; Plec; PLEC1; PLEC1b; Plectin; plectin 1; plectin 1, intermediate filament binding protein 500kDa; plectin-1; Plectin-6; Pltn | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18810, 5339, 64204 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.