missing translation for 'onlineSavingsMsg'
Learn More

PLEK Mouse anti-Human, Porcine, Clone: 1D5C2, Proteintech

Mouse Monoclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen PLEK
Clone 1D5C2
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16838305
View Documents
Proteintech
66431-1-IG-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16828305
View Documents
Proteintech
66431-1-IG-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Major protein kinase C substrate of platelets.
Specifications

Specifications

PLEK
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P47, Platelet 47 kDa protein, pleckstrin, PLEK
PLEK
IgG1
Protein G
Antibody
1D5C2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Pig
P08567
100516257, 5341
PLEK Fusion Protein Ag3187
Primary
-20°C
PLEK
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.