missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA3 Antibody (OTI5D11), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73470PCP
This item is not returnable.
View return policy
Description
PLEKHA3 Monoclonal specifically detects PLEKHA3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| PLEKHA3 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| PLEKHA3 | |
| Full length recombinant protein of human PLEKHA3 (NP_061964) produced in HEK293T cells. | |
| 0.1 mL | |
| 65977.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI5D11 | |
| Flow Cytometry | |
| FAPP-1, FAPP1PH domain-containing family A member 3, FLJ20067, four-phosphate-adaptor protein 1, Phosphatidylinositol-four-phosphate adapter protein 1, Phosphoinositol 4-phosphate adapter protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 3, pleckstrin homology domain-containing family A member 3, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction