Learn More
Invitrogen™ PLEKHA3 Monoclonal Antibody (OTI5D11)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525549
Product Code. 15915154
  • € 572.00 / 100µL
Description
PLEKHA3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
PLEKHA3 (Pleckstrin Homology Domain Containing A3) is a Protein Coding gene. Among its related pathways are Metabolism and Cholesterol and Sphingolipids transport / Transport from Golgi and ER to the apical membrane (normal and CF). Gene Ontology (GO) annotations related to this gene include phospholipid binding and 1-phosphatidylinositol binding. An important paralog of this gene is PLEKHA8.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLEKHA3
Monoclonal
1.1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HB20
Plekha3
Full length human recombinant protein of PLEKHA3 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI5D11
Unconjugated
Plekha3
FAPP1; FAPP-1; four-phosphate-adaptor protein 1; PH domain-containing family A member 3; Phosphatidylinositol-four-phosphate adapter protein 1; phosphoinositol 4-phosphate adapter protein 1; pleckstrin homology domain containing A3; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 3; pleckstrin homology domain-containing family A member 3; pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3; Plekha3
Mouse
Affinity Chromatography
RUO
65977
-20°C, Avoid Freeze/Thaw Cycles