missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA3 Monoclonal Antibody (OTI6E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502876S
This item is not returnable.
View return policy
Description
PLEKHA3 Monoclonal antibody specifically detects PLEKHA3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| PLEKHA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HB20 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 65977 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI6E4 | |
| Unconjugated | |
| Plekha3 | |
| FAPP1; FAPP-1; four-phosphate-adaptor protein 1; PH domain-containing family A member 3; Phosphatidylinositol-four-phosphate adapter protein 1; phosphoinositol 4-phosphate adapter protein 1; pleckstrin homology domain containing A3; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 3; pleckstrin homology domain-containing family A member 3; pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3; Plekha3 | |
| Full length human recombinant protein of human PLEKHA3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction