Learn More
PLEKHF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20389-1-AP-20UL
This item is not returnable.
View return policy
Description
APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.Specifications
| PLEKHF1 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHF1 | |
| Apoptosis inducing protein, APPD, LAPF, Phafin 1, PHAFIN1, PLEKHF1, ZFYVE15 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 72287, 79156 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TB82, Q96S99 | |
| Plekhf1 | |
| PLEKHF1 Fusion Protein Ag14228 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |