Learn More
PLEKHF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20389-1-AP-20UL
 View more versions of this product
Product Code. 16879924
  • € 145.00 / 20µL
Description
APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.
Specifications
Specifications
PLEKHF1
Polyclonal
Unconjugated
PLEKHF1
Apoptosis inducing protein, APPD, LAPF, Phafin 1, PHAFIN1, PLEKHF1, ZFYVE15
Rabbit
Antigen Affinity Chromatography
RUO
72287, 79156
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3TB82, Q96S99
Plekhf1
PLEKHF1 Fusion Protein Ag14228
20 μL
Primary
Human, Mouse