missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHH2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06559APC
This item is not returnable.
View return policy
Description
PLEKHH2 Polyclonal antibody specifically detects PLEKHH2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| PLEKHH2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| KIAA2028, pleckstrin homology domain containing, family H (with MyTH4 domain) member 2, PLEKHH1L | |
| Produced in rabbits immunized with E. coli-derived Human PLEKHH2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 130271 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction