missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHH3 Antibody (4F8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079990-M03
This item is not returnable.
View return policy
Description
PLEKHH3 Monoclonal antibody specifically detects PLEKHH3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| PLEKHH3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| FLJ21019, PH domain-containing family H member 3, pleckstrin homology domain containing, family H (with MyTH4 domain) member 3, pleckstrin homology domain-containing family H member 3 | |
| PLEKHH3 (NP_079203, 665 a.a. ∽ 750 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DVLELSTEPGRGAPQKLCLGLGAKAMSLSRPGETEPIHSVSYGHVAACQLMGPHTLALRVGESQLLLQSPQVEEIMQLVNAYLANP | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4F8 | |
| Western Blot 1:500, ELISA | |
| NP_079203 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 79990 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction