missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHM2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77089AF488
This item is not returnable.
View return policy
Description
PLEKHM2 Polyclonal antibody specifically detects PLEKHM2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PLEKHM2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| KIAA0842RP11-169K16.1, novel RUN and PH domain-containing protein, PH domain-containing family M member 2, pleckstrin homology domain containing, family M (with RUN domain) member 2, pleckstrin homology domain-containing family M member 2, Salmonella-induced filaments A and kinesin-interacting protein, SifA (Salmonella-induced filaments A) and kinesin-interacting protein, SifA and kinesin-interacting protein, SKIP | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human PLEKHM2. The immunogen is located within the last 50 amino acids of PLEKHM2. Amino Acid Squence: TAIQEASNKKKFEDALS | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 23207 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction