missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLEKHM3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15785021 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15785021 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15785021 Supplier Invitrogen™ Supplier No. PA557127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MIWDHCKSRL LETKAQNVFP AKEQFMVQRG TTPDNLSWME QKEASTFNFF NICQRRRDRP RSVNDLLDET STFKPGHARS RSDITQVD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

PLEKHM3, also known as DAPR, is a member of the M family of Pleckstrin homology domain-containing proteins. PLEKHM3 was initially identified through chromatin immunoprecipitation and CpG microarray analysis examining proteins regulated by myocyte-enhancing factor 2. In C2C12 myoblast cells, PLEKHM3 binds to the PI3K signaling member protein kinase B in the cytosol prior to differentiation into myotubes. Following the initiation of differentiation, PLEKHM3 was also found in membrane fractions. Knockdown of PLEKHM3 expression by RNAi resulted in the inhibition of myotube formation, suggesting that PLEKHM3 is a key component required by myoblasts for orchestrating their differentiation during myogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHM3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHM3
Gene Accession No. Q6ZWE6, Q8BM47
Gene Alias 9430067K14Rik; A230102O09Rik; AI449570; Dapr; differentiation associated protein; Differentiation-associated protein; PH domain-containing family M member 3; pleckstrin homology domain containing M3; pleckstrin homology domain containing, family M, member 1-like; pleckstrin homology domain containing, family M, member 3; pleckstrin homology domain-containing family M member 3; PLEKHM1L; Plekhm3; RGD1307151
Gene Symbols PLEKHM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLEKHM3. Recombinant protein control fragment (Product #RP-96208).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241075, 316455, 389072
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.