missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin A1 Antibody (708954) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB65361B
This item is not returnable.
View return policy
Description
Plexin A1 Monoclonal antibody specifically detects Plexin A1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| Plexin A1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 708954 | |
| Flow Cytometry, Immunocytochemistry | |
| NOV, NOVP, plexin A1, PLEXIN-A1 | |
| E. coli-derived recombinant human Plexin A1, Tyr964-Leu1152, Accession # Q9UIW2 | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience | |
| 5361 | |
| Store at 4°C in the dark. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion