missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin A1 Antibody (708954), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB65361I
This item is not returnable.
View return policy
Description
Plexin A1 Monoclonal antibody specifically detects Plexin A1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| Plexin A1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 708954 | |
| Flow Cytometry, Immunocytochemistry | |
| NOV, NOVP, plexin A1, PLEXIN-A1 | |
| E. coli-derived recombinant human Plexin A1, Tyr964-Leu1152, Accession # Q9UIW2 | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience | |
| 5361 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction