missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin A1 Antibody (M535), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23316
This item is not returnable.
View return policy
Description
Plexin A1 Monoclonal antibody specifically detects Plexin A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| Plexin A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| M535 | |
| Western Blot 1:500, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:50 | |
| NOV, NOVP, plexin A1, PLEXIN-A1 | |
| Clone M535 was generated from a Plexin A1 synthetic peptide (coupled to carrier protein) corresponding to a region within the N-terminal half of the semaphorin domain in human Plexin A1. This sequence is highly conserved in rat and mouse Plexin A1, and has low homology to other Plexin family members. [UniProt# Q9UIW2] | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience | |
| 5361 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction