Learn More
Invitrogen™ Plexin-A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143746
Description
The antibody detects a 220 kDa* protein corresponding to the apparent molecular mass of Plexin-A1 on SDS-PAGE immunoblots of adult mouse brain and Cos-7 cells transfected with mouse Plexin-A1. In immunocytochemistry, this antibody co-localizes with anti-Myc in Cos-7 cells transfected with Myc-tagged mouse Plexin-A1.
Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt].
Specifications
| Plexin-A1 | |
| Polyclonal | |
| Unconjugated | |
| PLXNA1 | |
| 2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18844, 362398, 5361 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P70206, Q9UIW2 | |
| PLXNA1 | |
| Plexin-A1 synthetic peptide (coupled to carrier protein) corresponding to a region within the semaphorin domain in human Plexin-A1. This sequence is highly conserved in rat and mouse Plexin-A1, and has low homology to other Plexin family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.