missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23325
This item is not returnable.
View return policy
Description
Plexin B1 Polyclonal antibody specifically detects Plexin B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Plexin B1 | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:400 | |
| plexin B1, SEP | |
| A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal region of human Plexin B1. This sequence has high homology with similar regions in rat and mouse Plexin B1, and has low homology to other plexin family members. [UniProt# O43157] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 5364 | |
| Store at -20°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction