missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Plexin C1 Antibody (2H11C11), Novus Biologicals™
Mouse Monoclonal Antibody
€ 287.00 - € 511.00
Specifications
| Antigen | Plexin C1 |
|---|---|
| Clone | 2H11C11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30039303
|
Novus Biologicals™
NBP3-27143-0.1ml |
0.1 mL |
€ 511.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30035460
|
Novus Biologicals™
NBP3-27143-0.025ml |
0.025 mL |
€ 287.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plexin C1 Monoclonal antibody specifically detects Plexin C1 in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| Plexin C1 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Growth and Development, Neuronal Cell Markers | |
| PBS | |
| 10154 | |
| IgG1 | |
| Protein G purified |
| 2H11C11 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPRplexin-C1, Virus-encoded semaphorin protein receptor | |
| Purified recombinant fragment of human Plexin C1 (AA: extra 35-234) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title