missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin D1 Antibody (752829), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4160F
This item is not returnable.
View return policy
Description
Plexin D1 Monoclonal antibody specifically detects Plexin D1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Plexin D1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23129 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 752829 | |
| Flow Cytometry, CyTOF-ready | |
| KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1 | |
| Mouse myeloma cell line NS0-derived recombinant human Plexin D1, Leu47-Ala1271, Accession # Q9Y4D7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction