missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plfr Antibody (RM0308-7N31), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12437UV
This item is not returnable.
View return policy
Description
Plfr Monoclonal specifically detects Plfr in Mouse samples. It is validated for Western Blot.
Specifications
| Plfr | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0308-7N31 | |
| Western Blot | |
| PLF-RP, Prl7d1, PRP | |
| Mouse recombinant Plfr | |
| 0.1 mL | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction