missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLIN4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143175
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Specifications
| PLIN4 | |
| Polyclonal | |
| Unconjugated | |
| PLIN4 | |
| adipocyte protein S3-12; KIAA1881; perilipin 4; perilipin-4; PLIN4; S3-12 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 57435, 729359 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O88492, Q96Q06 | |
| PLIN4 | |
| Peptide with sequence C-QRAFEHALSHIQHN. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction