missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLIN5 Mouse anti-Human, Clone: OTI3F2, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811630S
This item is not returnable.
View return policy
Description
PLIN5 Monoclonal antibody specifically detects PLIN5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PLIN5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q00G26 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 440503 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F2 | |
| Unconjugated | |
| Plin5 | |
| 2310076L09Rik; AI415325; AW109675; lipid droplet associated protein; lipid droplet-associated protein PAT-1; Lipid storage droplet protein 5; LSDA5; LSDP5; Mldp; Myocardial LD protein; Oxpat; Pat1; PAT-1; perilipin 5; perilipin-5; Plin5 | |
| Human recombinant protein fragment corresponding to amino acids 1-463 of human PLIN5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction