missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody (AZ44), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2747AF647
This item is not returnable.
View return policy
Description
PLK1 Monoclonal specifically detects PLK1 in Xenopus samples. It is validated for Western Blot, ELISA.
Specifications
| PLK1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PLK1 | |
| Baculo-expressed PLK1 from Xenopus laevis | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| 5347 | |
| Xenopus species | |
| Purified |
| Western Blot, ELISA | |
| AZ44 | |
| Western Blot, ELISA | |
| cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Plx is activated by phosphorylation. AZ44 only detects the mitotic form of Plx (Xenopus laevis) which may be due to the recognition of a phosphorylated epitope. Recommended concentration: 1 ug/ml | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction