Learn More
Invitrogen™ PLK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597143
Description
Immunogen sequence: VLFNDGTHMA LSANRKTVHY NPTSTKHFSF SVGAVPRALQ PQLGILRYFA SYMEQHLMKG GDLPSVEEVE VPAPPLLLQW VKTDQALLML FSDGTVQVNF YGDHTKLILS GWEPLLVTFV ARNRSACTYL ASHLRQLGCS PDLRQRLRYA LRLLRDRSPA; Positive Samples: A-549, HT-1080, SKOV3, 293T, U2OS, Mouse liver, Rat liver; Cellular Location: Cytoplasm, Golgi apparatus, Nucleus, centrosome, cytoskeleton, microtubule organizing center, nucleolus.
PLK3 is a member of the polo family of serine/threonine kinases. PLK3 is involved in the regulation of M phase in the cell cycle. CNK contains both a catalytic domain and a putative regulatory domain. It may play a role in regulation of cell cycle progression and tumorigenesis.
Specifications
| PLK3 | |
| Polyclonal | |
| Unconjugated | |
| PLK3 | |
| CNK; cytokine inducible kinase; cytokine-inducible serine/threonine-protein kinase; FGF-inducible kinase; Fnk; PLK3; PLK-3; polo like kinase 3; polo-like kinase 3; PRK; Proliferation-related kinase; serine/threonine-protein kinase PLK3; serine-threonine kinase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1263, 12795, 58936 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q60806, Q9H4B4, Q9R011 | |
| PLK3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 487-646 of human PLK3 (NP_0040642). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.