missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK4 Monoclonal Antibody (OTI2A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810550S
This item is not returnable.
View return policy
Description
PLK4 Monoclonal antibody specifically detects PLK4 in Human samples. It is validated for Western BlotSpecifications
| PLK4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96ND8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 147949 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2A12 | |
| Unconjugated | |
| ZNF583 | |
| 1700028H20; AI385771; MCCRP2; PLK4; PLK-4; polo like kinase 4; polo-like kinase 4; SAK; serine/threonine kinase 18; serine/threonine-protein kinase 18; Serine/threonine-protein kinase PLK4; serine/threonine-protein kinase Sak; Snk akin kinase; Stk18 | |
| Full length human recombinant protein of human ZNF583 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction