missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93608-0.02ml
This item is not returnable.
View return policy
Description
PLOD1 Polyclonal antibody specifically detects PLOD1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PLOD1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| 2-oxoglutarate 5-dioxygenase 1, EC 1.14.11.4, Ehlers-Danlos syndrome type VI), FLJ42041, LH1, LLH, lysine hydroxylase, Lysyl hydroxylase 1, PLODLH, procollagen-lysine, procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase (lysine hydroxylase, procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1, procollagen-lysine, 2-oxoglutarate 5-dioxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-295 of human PLOD1 (NP_000293.2). KGDAKPEDNLLVLTVATKETEGFRRFKRSAQFFNYKIQALGLGEDWNVEKGTSAGGGQKVRLLKKALEKHADKEDLVILFADSYDVLFASGPRELLKKFRQARSQVVFSAEELIYPDRRLETKYPVVSDGKRFLGSGGFIGYAPNLSKLVAEWEGQDSDSDQLFYTKIFLDPEKREQINITLDHRCRIFQNLDGALDEVVLKFEMGHVRARNLAYDTLPVLIHGNGPTKLQLNYLGNYIPRFWTFETGCTVCDEGLRSLKGIGDEALPTVLVGVFIE | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 5351 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction