missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33264-100ul
This item is not returnable.
View return policy
Description
PLOD3 Monoclonal antibody specifically detects PLOD3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PLOD3 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| 2-oxoglutarate 5-dioxygenase 3, EC 1.14.11.4, LH3Lysyl hydroxylase 3, lysine hydroxylase 3, procollagen-lysine, procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-306 of human PLOD3 (NP_001075.1).,, Sequence:, SDRPRGRDPVNPEKLLVITVATAETEGYLRFLRSAEFFNYTVRTLGLGEEWRGGDVARTVGGGQKVRWLKKEMEKYADREDMIIMFVDSYDVILAGSPTELLKKFVQSGSRLLFSAESFCWPEWGLAEQYPEVGTGKRFLNSGGFIGFATTIHQIVRQWKYKDDDDDQLFYTRLYLDPGLREKLSLNLDHKSRIFQNLNGALDEVVLKFDRNRVRIRNVAYDTLPIVVHGNGPTKLQLNYLGNYVPNGWTPEGGCGFCNQDRRTLPGGQPPPRVFLAVFVEQ | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8985 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction