Learn More
Invitrogen™ PLOD3 Recombinant Rabbit Monoclonal Antibody (5R6Q9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551264
Description
Immunogen sequence: SDRPRGRDPV NPEKLLVITV ATAETEGYLR FLRSAEFFNY TVRTLGLGEE WRGGDVARTV GGGQKVRWLK KEMEKYADRE DMIIMFVDSY DVILAGSPTE LLKKFVQSGS RLLFSAESFC WPEWGLAEQY PEVGTGKRFL NSGGFIGFAT TIHQIVRQWK YKDDDDDQLF YTRLYLDPGL REKLSLNLDH KSRIFQNLNG ALDEVVLKFD RNRVRIRNVA YDTLPIVVHG NGPTKLQLNY LGNYVPNGWT PEGGCGFCNQ DRRTLPGGQP PPRVFLAVFV EQ.
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
Specifications
| PLOD3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O60568, Q9R0E1 | |
| PLOD3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-306 of human PLOD3 (NP_001075.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 5R6Q9 | |
| Unconjugated | |
| PLOD3 | |
| AI414586; bone protein I (BP-I); Galactosylhydroxylysine-glucosyltransferase; LH3; lysine hydroxylase 3; lysyl hydroxlase 3; lysyl hydroxylase 2; lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; Plod3; Procollagen galactosyltransferase; Procollagen glucosyltransferase; Procollagen glycosyltransferase; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3; procollagen-lysine,2-oxoglutarate 5-dioxygenase 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26433, 8985 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.