missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLPP4/PPAPDC1A Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81931DL594
This item is not returnable.
View return policy
Description
PLPP4/PPAPDC1A Polyclonal antibody specifically detects PLPP4/PPAPDC1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PLPP4/PPAPDC1A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 594 | |
| Diacylglycerol Pyrophosphate Like 2, DPPL2, Phosphatidate Phosphatase PPAPDC1A, Phosphatidic Acid Phosphatase Type 2 Domain Containing 1A, Phospholipid Phosphatase 4, PPAPDC1 | |
| PPAPDC1A antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PPAPDC1A. The immunogen is located within the last 50 amino acids of PPAPDC1A. Amino Acid Squence: ASLKKEERPTADSA | |
| 0.1 mL | |
| Signal Transduction | |
| 196051 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction