missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLSCR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-36713-100ul
This item is not returnable.
View return policy
Description
PLSCR3 Polyclonal antibody specifically detects PLSCR3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| PLSCR3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Ca(2+)-dependent phospholipid scramblase 3, phospholipid scramblase 3, PL scramblase 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-295 of human PLSCR3 (NP_065093.2).,, Sequence:, MAGYLPPKGYAPSPPPPYPVTPGYPEPALHPGPGQAPVPAQVPAPAPGFALFPSPGPVALGSAAPFLPLPGVPSGLEFLVQIDQILIHQKAERVETFLGWETCNRYELRSGAGQPLGQAAEESNCCARLCCGARRPLRVRLADPGDREVLRLLRPLHCGCSCCPCGLQEMEVQAPPGTTIGHVLQTWHPFLPKFSIQDADRQTVLRVVGPCWTCGCGTDTNFEVKTRDESRSVGRISKQWGGLVREALTDADDFGLQFPLDLDVRVKAVLLGATFLIDYMFFEKRGGAGPSAVTS | |
| 100 μL | |
| metabolism | |
| 57048 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction