missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLSCR3 Recombinant Rabbit Monoclonal Antibody (4V2P5)

Product Code. 30131781
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30131781 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131781 Supplier Invitrogen™ Supplier No. MA555368

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: MAGYLPPKGY APSPPPPYPV TPGYPEPALH PGPGQAPVPA QVPAPAPGFA LFPSPGPVAL GSAAPFLPLP GVPSGLEFLV QIDQILIHQK AERVETFLGW.

May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system. Seems to play a role in apoptosis, through translocation of cardiolipin from the inner to the outer mitochondrial membrane which promotes BID recruitment and enhances tBid-induced mitochondrial damages. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLSCR3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4V2P5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Plscr3
Gene Accession No. Q6QBQ4, Q9JIZ9, Q9NRY6
Gene Alias 2210403O21Rik; 2610037N06Rik; Ca 2+ dependent phospholipid scramblase 3; ca(2+)-dependent phospholipid scramblase 3; ESTM3; LOW QUALITY PROTEIN: phospholipid scramblase 3; LOW QUALITY PROTEIN: phospholipid scramblase 3; phospholipid scramblase 3; phospholipid scramblase 3; PL scramblase 3; Pls3; Plscr3; Unknown (protein for MGC:138060); X83310
Gene Symbols Plscr3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PLSCR3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360549, 57048, 70310
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.