missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLSCR4 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97270IR
This item is not returnable.
View return policy
Description
PLSCR4 Polyclonal antibody specifically detects PLSCR4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PLSCR4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| Ca(2+)-dependent phospholipid scramblase 4, cell growth inhibiting protein 43, Cell growth-inhibiting gene 43 protein, phospholipid scramblase 4, PL scramblase 4, TRA1 | |
| Produced in rabbits immunized with E. coli-derived Human PLSCR4 fragment. | |
| 0.1 mL | |
| Cardiovascular Biology, Neuroscience, Signal Transduction | |
| 57088 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction