missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLTP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA132254
This item is not returnable.
View return policy
Description
PA1-32254 detects PLTP from mouse, human samples.
PLTP is one of at least two lipid transfer proteins found in human plasma. The protein transfers phospholipids from triglyceride-rich lipoproteins to high density lipoprotein (HDL). In addition to regulating the size of HDL particles, this protein may be involved in cholesterol metabolism.
Specifications
| PLTP | |
| Polyclonal | |
| Unconjugated | |
| PLTP | |
| BPI fold containing family E; BPIFE; HDLCQ9; lipid transfer protein II; OD107; Phospholipid transfer protein; Pltp | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| whole serum with 0.02% sodium azide | |
| P55058, P55065 | |
| PLTP | |
| A partial peptide of human PLTP. | |
| RUO | |
| 18830, 5360 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction