Learn More
SI Analytics™ Connection Cables and Plugs
Product Code. 10535553 Shop All SI Analytics Products
Click to view available options
Length (Metric) Cable:
1 m
10 m
2 m
5 m
Type:
K 1 A
K 10 A
K2X
K5A
L 1 A
L 1 BNC
L 1 N
L 1 NN
L 1 R
L 2 A
Plug Cable Combination
Unit Size:
Each
Product Code. 10535553
Brand: SI Analytics™ 285120303
Only null left
Add to basket
Add to basket
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifications
Connector Types DIN
Type K 10 A
Includes 10m Coaxial Cable, DIN Instrument Plug
Length (Metric) Cable 5 m