missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Plug-In Tube Extension for IN450plus, Straight

Product Code. 17597745 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN450plus Incubator
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17597745

Brand: Memmert™ IN450plusB29719

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.

  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight

Specifications

For Use With (Equipment) IN450plus Incubator
Type Plug-in Tube Extension
Diameter (Metric) O.D. 60.3 mm, I.D. 57 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.