Learn More
Memmert™ Plug-In Tube Extension for IN55, Straight
Product Code. 17587795 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IN55 Incubator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17587795 IN55 Incubator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17587795 Supplier Memmert™ Supplier No. IN55B29718
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
For Use With (Equipment) IN55 Incubator
Type Plug-in Tube Extension
Diameter (Metric) O.D. 60.3 mm, I.D. 57 mm