Learn More
Memmert™ Plug-In Tube Extension for SF30, Straight
Product Code. 17538215 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SF30 Sterilizer
Unit Size:
Each
Product Code. 17538215
Brand: Memmert™ SF30B29719
Only null left
Add to basket
Add to basket
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Diameter (Metric) Outside 60.3 mm
For Use With (Equipment) SF30 Sterilizer