Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
Specifications
| Diameter (Metric) Outside | 60.3 mm |
| For Use With (Equipment) | SN260plus Sterilizer |