missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLUNC Monoclonal Antibody (251512)
GREENER_CHOICE

Product Code. 15523642
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15523642 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15523642 Supplier Invitrogen™ Supplier No. MA523949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

PLUNC is the human homolog of murine plunc and is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this protein is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. This gene is the human homolog of murine plunc, and like the mouse gene, is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this gene is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. Multiple transcript variants resulting from alternative splicing in the 3' UTR have been detected, but the full-length nature of only two is known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLUNC
Applications Western Blot
Classification Monoclonal
Clone 251512
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Bpifa1
Gene Accession No. Q9NP55
Gene Alias bA49G10.5; BPI fold containing family A member 1; BPI fold containing family A member 1 protein; BPI fold containing family A, member 1; BPI fold-containing family A member 1; BPIFA1; ES cell-related protein; ligand-binding protein RYA3; LPLUNC3; lung-specific protein X; LUNX; LUNX-like protein; LUNXNASG; NASG; Nasopharyngeal carcinoma-related protein; palate lung and nasal epithelium clone protein; palate, lung and nasal epithelium associated; palate, lung and nasal epithelium carcinoma associated; palate, lung, and nasal epithelium associated; palate, lung, and nasal epithelium carcinoma associated protein; palate, lung, and nasal epithelium clone; palate, lung, and nasal epithelium expressed transcript; PLUNC; protein Plunc; secretory protein in upper respiratory tracts; short palate; Short PLUNC1; SPLUNC1; SPLUNC1SPURT; SPURT; tracheal epithelium enriched protein; tracheal epithelium-enriched protein; UNQ787/PRO1606; Von Ebner protein Hl
Gene Symbols Bpifa1
Host Species Mouse
Immunogen E. coli-derived recombinant human PLUNC Gln20-Val256.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51297
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.