Learn More
Trajan Scientific and Medical™ Replacement Plunger for Fixed Needle Thermo RSH Autosampler Syringes
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Unit Size:
Each
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
Specifications
Specifications
| Material | PTFE (Plunger) |
| Product Type | Gastight |
| Volume (Metric) | 500 μL |