Learn More
Trajan Scientific and Medical™ Plunger Replacement For Fixed Needle Headspace Syringe
Click to view available options
Volume (Metric):
1 mL
2.5 mL
Unit Size:
Each
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
- For use with Thermo Scientific RSH Autosamplers
Specifications
Specifications
| Material | PTFE (Plunger) |
| Volume (Metric) | 2.5 mL |
| For Use With (Equipment) | Thermo RSH Autosampler |
| Connection Type | Gas Tight |