missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLX647, MedChemExpress
Description
PLX647 is an orally active, highly specific dual FMS and KIT kinase inhibitor, with IC50s of 28 and 16 nM, respectively. PLX647 shows selectivity for FMS and KIT over a panel of 400 kinases at a concentration of 1 μM except FLT3 and KDR (IC50s=91 and 130 nM, respectively).
Specifications
Specifications
| Chemical Name or Material | PLX647 |
| CAS | 873786-09-5 |
| Color | Deep Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H17F3N4 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 25 mg/mL (65.38 mM; Need ultrasonic) |
| SMILES | FC(C1=CC=C(CNC2=NC=C(CC3=CNC4=NC=CC=C43)C=C2)C=C1)(F)F |
| Molecular Weight (g/mol) | 382.38 |
| Formula Weight | 382.38 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?