Learn More
Novus Biologicals™ Plxdc2 Antibody (Fox Chase patent anti-TEM7R), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28803-100ug
 View more versions of this product
Product Code. 30045070
  • € 338.00 / 100µg
Description
Plxdc2 Monoclonal antibody specifically detects Plxdc2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plxdc2
Monoclonal
Unconjugated
Q6UX71
Human
Protein A purified
RUO
84898
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Fox Chase patent anti-TEM7R
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
plexin domain containing 2,1200007L24Rik, plexin domain-containing protein 2, TEM7RFLJ14623, Tumor endothelial marker 7-related protein
TEM7R/ PLXDC2
100 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.