Learn More
Novus Biologicals™ Plxdc2 Antibody (Fox Chase patent anti-TEM7R), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28803-1mg
This item is not returnable.
View return policy
Description
Plxdc2 Monoclonal antibody specifically detects Plxdc2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Plxdc2 | |
| Monoclonal | |
| Unconjugated | |
| Q6UX71 | |
| Human | |
| Protein A purified | |
| RUO | |
| 84898 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Fox Chase patent anti-TEM7R | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| plexin domain containing 2,1200007L24Rik, plexin domain-containing protein 2, TEM7RFLJ14623, Tumor endothelial marker 7-related protein | |
| TEM7R/ PLXDC2 | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |