missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXDC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12285-1-AP-20UL
This item is not returnable.
View return policy
Description
May play a role in tumor angiogenesis.Specifications
| PLXDC2 | |
| Polyclonal | |
| Unconjugated | |
| PLXDC2 | |
| plexin domain containing 2, PLXDC2, TEM7R | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361282, 67448, 84898 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6UX71, Q9DC11 | |
| Plxdc2 | |
| PLXDC2 Fusion Protein Ag2940 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction