missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLXNA3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15558062
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15558062 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558062 Supplier Invitrogen™ Supplier No. PA547571

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A2, recombinant human (rh) Plexin B1, rhPlexin C1, and rhPlexin D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Coreceptor for SEMA3A and SEMA3F. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Regulates the migration of sympathetic neurons, but not of neural crest precursors. Required for normal dendrite spine morphology in pyramidal neurons. May play a role in regulating semaphorin-mediated programmed cell death in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNA3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Plxna3
Gene Accession No. D3ZPX4, P70208
Gene Alias 6.3; HSSEXGENE; mKIAA4078; PlexA3; plexin 3; Plexin 4, SEX, homolog to the cMet/HGF receptors; plexin A3; plexin-4; Plexin-A3; Plxa3; Plxn3; Plxn4; Plxna3; Semaphorin receptor SEX; SEX; Sex chromosome X transmembrane protein of HGF receptor family 3; SEXPLEXIN-A3; XAP-6
Gene Symbols Plxna3
Host Species Goat
Immunogen E. coli-derived recombinant mouse Plexin A3 Leu34-Asp150.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18846, 309280
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.