Learn More
Invitrogen™ PLXNB1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5143747
Product Code. 17857058
  • € 546.00 / 100µL
Description
The antibody detects 300, 150, and 100 kDa* bands on SDS-PAGE immunoblots of mouse liver and rat PC12 cells. These bands correspond to the full length and proteolytically processed forms of Plexin B1. In mouse liver, this antibody detects 300 and 100 kDa forms of Plexin B1.
Plexin B1 is a protein of the plexin family that in humans is encoded by the PLXNB1gene. Receptor for SEMA4D. Plays a role in RHOA activation and subsequent changes of the actin cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLXNB1
Polyclonal
Unconjugated
Plxnb1
2900002G15Rik; AU042020; Kiaa0407; mKIAA0407; plexin 1; plexin 5; plexin B1; plexin-A1; PLEXIN-B1; PLXN5; Plxnb1; Semaphorin receptor SEP; SEP
Rabbit
Antigen Affinity Chromatography
RUO
235611, 316009, 5364
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1.0 mg/mL
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
O43157, Q8CJH3
Plxnb1
A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal region of human Plexin B1. This sequence has high homology with similar regions in rat and mouse Plexin B1, and has low homology to other plexin family members.
100 μL
Primary
Human, Mouse, Rat